Print…
Anti-PIWIL4 Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111714-50EA 998 EUR
ABCAAB111714-50
Anti-PIWIL4 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: PIWIL4 (piwi-like RNA-mediated gene silencing 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human