Print…
Anti-ANXA8 Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111708-50EA 993 EUR
ABCAAB111708-50
Anti-ANXA8 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: ANXA8 (annexin A8)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human