Print…
Anti-E2 230K Rabbit polyclonal antibody unconjugated
ABCAAB111707-50EA 698 EUR
ABCAAB111707-50
Anti-E2 230K Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: E2 230K
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human