Print…
Anti-SNX18 Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111702-50EA 883 EUR
ABCAAB111702-50
Anti-SNX18 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: SNX18 (sorting nexin 18)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human