Print…
Anti-UPRT Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111701-50EA 921 EUR
ABCAAB111701-50
Anti-UPRT Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: UPRT (uracil phosphoribosyltransferase (FUR1) homolog (S. cerevisiae))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human