Print…
Anti-ZNF587 Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111697-50EA 921 EUR
ABCAAB111697-50
Anti-ZNF587 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: ZNF587 (zinc finger protein 587)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human