Print…
Anti-TMPRSS5 Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB111695-50EA 921 EUR
ABCAAB111695-50
Anti-TMPRSS5 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: TMPRSS5 (transmembrane protease, serine 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human