Print…
Anti-IGF1 Goat Polyclonal Antibody
Fornecedor: Abcam
ABCAAB106836-100EA 971 EUR
ABCAAB106836-100
Anti-IGF1 Goat Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: IGF1 (insulin-like growth factor 1 (somatomedin C))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human