Print…
Anti-NANOG Chicken polyclonal antibody unconjugated
ABCAAB90168-100EA 740 EUR
ABCAAB90168-100
Anti-NANOG Chicken polyclonal antibody unconjugated
Type: Primary
Antigen: NANOG (Nanog homeobox)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human