Print…
Anti-PLAUR Mouse Monoclonal Antibody [clone: MM0597-18L1]
Fornecedor: Abcam
ABCAAB89932-50EA 759 EUR
ABCAAB89932-50
Anti-PLAUR Mouse Monoclonal Antibody [clone: MM0597-18L1]
Anticorpos
Type: Primary
Antigen: PLAUR (plasminogen activator, urokinase receptor)
Clonality: monoclonal
Clone: MM0597-18L1
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human