Print…
Anti-ULBP2 Mouse Monoclonal Antibody [clone: MM0593-7F33]
Fornecedor: Abcam
ABCAAB89930-50EA 965 EUR
ABCAAB89930-50
Anti-ULBP2 Mouse Monoclonal Antibody [clone: MM0593-7F33]
Anticorpos
Type: Primary
Antigen: ULBP2 (UL16 binding protein 2)
Clonality: monoclonal
Clone: MM0593-7F33
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human