Print…
Anti-RELT Mouse Monoclonal Antibody [clone: MM0527-8G28]
Fornecedor: Abcam
ABCAAB89922-50EA 964 EUR
ABCAAB89922-50
Anti-RELT Mouse Monoclonal Antibody [clone: MM0527-8G28]
Anticorpos
Type: Primary
Antigen: RELT (RELT tumor necrosis factor receptor)
Clonality: monoclonal
Clone: MM0527-8G28
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human