Print…
Anti-RLN3 Mouse Monoclonal Antibody [clone: MM0526-7G14]
Fornecedor: Abcam
ABCAAB89920-50EA 723 EUR
ABCAAB89920-50
Anti-RLN3 Mouse Monoclonal Antibody [clone: MM0526-7G14]
Anticorpos
Type: Primary
Antigen: RLN3 (relaxin 3)
Clonality: monoclonal
Clone: MM0526-7G14
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human