Print…
Anti-RLN1 Rat monoclonal antibody unconjugated [clone: MM0525-6L12]
ABCAAB89919-50EA 733 EUR
ABCAAB89919-50
Anti-RLN1 Rat monoclonal antibody unconjugated [clone: MM0525-6L12]
Type: Primary
Antigen: RLN1 (relaxin 1)
Clonality: monoclonal
Clone: MM0525-6L12
Conjugation: unconjugated
Epitope:
Host: Rat
Isotype: IgG2
Reactivity: Human