Print…
Anti-AGER Mouse Monoclonal Antibody [clone: MM0520-8D11]
Fornecedor: Abcam
ABCAAB89911-100EA 942 EUR
ABCAAB89911-100
Anti-AGER Mouse Monoclonal Antibody [clone: MM0520-8D11]
Anticorpos
Type: Primary
Antigen: AGER (advanced glycosylation end product-specific receptor)
Clonality: monoclonal
Clone: MM0520-8D11
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human