Print…
Anti-PSEN2 Mouse Monoclonal Antibody [clone: MM0516-7F8]
Fornecedor: Abcam
ABCAAB89910-50EA 759 EUR
ABCAAB89910-50
Anti-PSEN2 Mouse Monoclonal Antibody [clone: MM0516-7F8]
Anticorpos
Type: Primary
Antigen: PSEN2 (presenilin 2)
Clonality: monoclonal
Clone: MM0516-7F8
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human