Print…
Anti-LAYN Mouse monoclonal antibody unconjugated [clone: MM0448-4F33]
ABCAAB89790-50EA 698 EUR
ABCAAB89790-50
Anti-LAYN Mouse monoclonal antibody unconjugated [clone: MM0448-4F33]
Type: Primary
Antigen: LAYN (layilin)
Clonality: monoclonal
Clone: MM0448-4F33
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human