Print…
Anti-GRP78 BiP Chicken polyclonal antibody unconjugated
ABCAAB89789-100EA 723 EUR
ABCAAB89789-100
Anti-GRP78 BiP Chicken polyclonal antibody unconjugated
Type: Primary
Antigen: GRP78 BiP
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgG
Reactivity: Human, Mouse, Rat