Print…
Anti-FDXR Mouse polyclonal antibody unconjugated
Fornecedor: Abcam
ABCAAB89785-50EA 732 EUR
ABCAAB89785-50
Anti-FDXR Mouse polyclonal antibody unconjugated
Anticorpos
Type: Primary
Antigen: FDXR (ferredoxin reductase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human