Print…

Anti-MAG Mouse Monoclonal Antibody

Fornecedor: Abcam
ABCAAB89780-100EA 1320 EUR
ABCAAB89780-100
Anti-MAG Mouse Monoclonal Antibody
Anticorpos
Type: Primary
Antigen: MAG (myelin associated glycoprotein)
Clonality: monoclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR