Print…
Anti-CAPN1 Mouse Polyclonal Antibody
Fornecedor: Abcam
ABCAAB89778-50EA 791 EUR
ABCAAB89778-50
Anti-CAPN1 Mouse Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: CAPN1 (calpain 1, (mu/I) large subunit)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human