Print…
Anti-Cathelicidin Mouse Polyclonal Antibody
Fornecedor: Abcam
ABCAAB89774-50EA 774 EUR
ABCAAB89774-50
Anti-Cathelicidin Mouse Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: Cathelicidin
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human