Print…
Anti-Lysosomal acid lipase Mouse Polyclonal Antibody
Fornecedor: Abcam
ABCAAB89771-50EA 971 EUR
ABCAAB89771-50
Anti-Lysosomal acid lipase Mouse Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: Lysosomal acid lipase
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human