Print…
Anti-MAPT Mouse Monoclonal Antibody [clone: 3G3]
Fornecedor: Abcam
ABCAAB89748-100EA 964 EUR
ABCAAB89748-100
Anti-MAPT Mouse Monoclonal Antibody [clone: 3G3]
Anticorpos
Type: Primary
Antigen: MAPT (microtubule-associated protein tau)
Clonality: monoclonal
Clone: 3G3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human