Print…
Anti-KLK11 Mouse monoclonal antibody unconjugated [clone: MM0427-4N39]
ABCAAB89664-50EA 698 EUR
ABCAAB89664-50
Anti-KLK11 Mouse monoclonal antibody unconjugated [clone: MM0427-4N39]
Type: Primary
Antigen: KLK11 (kallikrein-related peptidase 11)
Clonality: monoclonal
Clone: MM0427-4N39
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2
Reactivity: Human