Print…
Anti-ANKRD28 Rabbit polyclonal antibody unconjugated
ABCAAB89657-200EA 733 EUR
ABCAAB89657-200
Anti-ANKRD28 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: ANKRD28 (ankyrin repeat domain 28)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human