Print…

Anti-FEZ2 Rabbit polyclonal antibody unconjugated

ABCAAB67410-100EA 698 EUR
ABCAAB67410-100
Anti-FEZ2 Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: FEZ2 (fasciculation and elongation protein zeta 2 (zygin II))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR