Print…

Anti-MX2 Mouse Polyclonal Antibody

Fornecedor: Abcam
ABCAAB67388-50EA 973 EUR
ABCAAB67388-50
Anti-MX2 Mouse Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: MX2 (myxovirus (influenza virus) resistance 2 (mouse))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR