Print…

Anti-CECR1 Mouse polyclonal antibody unconjugated

Fornecedor: Abcam
ABCAAB67380-50EA 779 EUR
ABCAAB67380-50
Anti-CECR1 Mouse polyclonal antibody unconjugated
Anticorpos
Type: Primary
Antigen: CECR1 (cat eye syndrome chromosome region, candidate 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR