Print…
Anti-Sodium / Hydrogen Exchanger 1 Rabbit Polyclonal Antibody
Fornecedor: Abcam
ABCAAB67313-100EA 997 EUR
ABCAAB67313-100
Anti-Sodium / Hydrogen Exchanger 1 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: Sodium / Hydrogen Exchanger 1
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat