Print…
Anti-CAMK2A Rabbit polyclonal antibody unconjugated
ABCAAB50202-100EA 751 EUR
ABCAAB50202-100
Anti-CAMK2A Rabbit polyclonal antibody unconjugated
Type: Primary
Antigen: CAMK2A (calcium/calmodulin-dependent protein kinase II alpha)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken, Human, Mouse, Rat