Print…
Anti-ATMPK6 Goat polyclonal antibody
ABCAAB50186-100EA 651 EUR
ABCAAB50186-100
Anti-ATMPK6 Goat polyclonal antibody
Type: Primary
Antigen: ATMPK6
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity:
Encomendar