Print…
Anti-Spinophilin/Neurabin 2 Rabbit polyclonal antibody
ABCAAB50184-100EA 684 EUR
ABCAAB50184-100
Anti-Spinophilin/Neurabin 2 Rabbit polyclonal antibody
Type: Primary
Antigen: Spinophilin/Neurabin 2
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat