Print…

Anti-TNFRSF6B Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB74377-100EA 0 EUR
BIRBORB74377-100
Anti-TNFRSF6B Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: TNFRSF6B (tumor necrosis factor receptor superfamily, member 6b, decoy)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR