Print…

Anti-IL1RAP Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB74374-100EA 0 EUR
BIRBORB74374-100
Anti-IL1RAP Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: IL1RAP (interleukin 1 receptor accessory protein)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR