Print…

Anti-CX3CR1 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB74362-100EA 0 EUR
BIRBORB74362-100
Anti-CX3CR1 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: CX3CR1 (chemokine (C-X3-C motif) receptor 1)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR