Print…

Anti-MAP3K14 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB74323-100EA 0 EUR
BIRBORB74323-100
Anti-MAP3K14 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: MAP3K14 (mitogen-activated protein kinase kinase kinase 14)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR