Print…

Anti-CXCR4 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB74308-100EA 0 EUR
BIRBORB74308-100
Anti-CXCR4 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: CXCR4 (chemokine (C-X-C motif) receptor 4)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR