Print…

Anti-UBA5 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB128886-200EA 0 EUR
BIRBORB128886-200 BIRBORB128886-100
Anti-UBA5 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: UBA5 (ubiquitin-like modifier activating enzyme 5)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Human, Mouse, Rat
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR