Print…
Anti-CHAF1B Rabbit Polyclonal Antibody
Fornecedor: Biorbyt
BIRBORB126808-100EA 0 EUR
BIRBORB126808-100 BIRBORB126808-200
Anti-CHAF1B Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: CHAF1B (chromatin assembly factor 1, subunit B (p60))
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar