Print…
Anti-CEP135 Rabbit Polyclonal Antibody
Fornecedor: Biorbyt
BIRBORB126804-100EA 0 EUR
BIRBORB126804-100 BIRBORB126804-200
Anti-CEP135 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: CEP135 (centrosomal protein 135kDa)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar