Print…
Anti-CEBPB Rabbit Polyclonal Antibody
Fornecedor: Biorbyt
BIRBORB126803-100EA 0 EUR
BIRBORB126803-100 BIRBORB126803-200
Anti-CEBPB Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: CEBPB (CCAAT/enhancer binding protein (C/EBP), beta)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar