Print…

Anti-CDH24 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB126792-100EA 0 EUR
BIRBORB126792-100 BIRBORB126792-200
Anti-CDH24 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: CDH24 (cadherin 24, type 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR