Print…

Anti-CCDC102A Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB126781-200EA 0 EUR
BIRBORB126781-200 BIRBORB126781-100
Anti-CCDC102A Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: CCDC102A (coiled-coil domain containing 102A)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR