Print…

Anti-CAD Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB126774-200EA 0 EUR
BIRBORB126774-200 BIRBORB126774-100
Anti-CAD Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: CAD (carbamoyl-phosphate synthetase 2, aspartate transcarbamylase, and dihydroorotase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR