Print…

Anti-C9orf89 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB126770-200EA 0 EUR
BIRBORB126770-200 BIRBORB126770-100
Anti-C9orf89 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: C9orf89 (chromosome 9 open reading frame 89)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR