Print…

Anti-C1QL2 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB126767-100EA 0 EUR
BIRBORB126767-100 BIRBORB126767-200
Anti-C1QL2 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: C1QL2 (complement component 1, q subcomponent-like 2)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR