Print…

Anti-BTBD6 Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB126763-100EA 0 EUR
BIRBORB126763-100 BIRBORB126763-200
Anti-BTBD6 Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: BTBD6 (BTB (POZ) domain containing 6)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR