Print…

Anti-BRI3B Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB126761-100EA 0 EUR
BIRBORB126761-100 BIRBORB126761-200
Anti-BRI3B Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: BRI3B
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR