Print…

Anti-BMX Rabbit Polyclonal Antibody

Fornecedor: Biorbyt
BIRBORB126756-200EA 0 EUR
BIRBORB126756-200 BIRBORB126756-100
Anti-BMX Rabbit Polyclonal Antibody
Anticorpos
Type: Primary
Antigen: BMX (BMX non-receptor tyrosine kinase)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity:
Encomendar

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mais info About VWR